Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD72 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114536 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-536 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-536 Supplier Abnova Corporation Supplier No. PAB0988

Rabbit polyclonal antibody raised against synthetic peptide of CD72.

Sequence: KDWKLTDDTQRTRI

Specifications

Antigen CD72
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description CD72 molecule
Formulation In PBS (0.02% sodium azide)
Gene CD72
Gene Alias CD72b, LYB2
Gene Symbols CD72
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 305-317 of human CD72.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 971
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.