Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Rabbit anti-CD79b, Polyclonal,
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ ABF187
Description
Specifically detects CD79b in Human samples, and it is validated for Immunoprecipitation, Western Blotting
The B-cell antigen receptor (BCR) is a multimeric complex consisting of an antigen-recognition structure and a membrane-bound immunoglobulin (mIg), associated non-covalently with a heterodimer of Ig-alpha (encoded by the CD79A gene) and Ig-beta (encoded by the CD79B gene). Signalling through the BCR is mediated by Ig-alpha and Ig-beta cytoplsmic domain.
Specifications
CD79b | |
Polyclonal | |
P40259 | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
Human | |
Serum |
Immunoprecipitation, Western Blot | |
Rabbit polyclonal serum with 0.05% sodium azide. | |
CD79B; B29; IGB | |
GST-tagged recombinant protein corresponding to the cytoplasmic domain of human CD79b. | |
100 μL | |
Inflammation and Immunology | |
NP_000617 | |
Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction