Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd97, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114684 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-684 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-684 Supplier Abnova Corporation Supplier No. PAB10186
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Cd97.

Specifications

Antigen Cd97
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Cd97.
Dilution Western Blot (1:1000-1:5000) Immunofluorescence (1:1000-1:5000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene Cd97
Gene Accession No. NP_036055;Q9Z0M6
Gene Alias AA409984/TM7LN1
Gene Symbols Cd97
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 1-512 of mouse Cd97.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26364
Target Species Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.