Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC2, Rabbit, Clone: SP80, Abnova™

Catalog No. 89108349 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-349 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-349 Supplier Abnova Corporation Supplier No. MAB10034

Rabbit monoclonal antibody raised against synthetic peptide of CDC2.

The protein encoded by this gene is a member of the Ser/Thr protein kinase family. This protein is a catalytic subunit of the highly conserved protein kinase complex known as M-phase promoting factor (MPF), which is essential for G1/S and G2/M phase transitions of eukaryotic cell cycle. Mitotic cyclins stably associate with this protein and function as regulatory subunits. The kinase activity of this protein is controlled by cyclin accumulation and destruction through the cell cycle. The phosphorylation and dephosphorylation of this protein also play important regulatory roles in cell cycle control. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen CDC2
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone SP80
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of CDC2.
Dilution Western Blot (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene CDC2
Gene Alias CDC28A/CDK1/DKFZp686L20222/MGC111195
Gene Symbols CDC2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human CDC2.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 983
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.