Learn More
CDC2 (phospho S39), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CDC2.
Supplier: Abnova Corporation PAB0436
Description
The protein encoded by this gene is a member of the Ser/Thr protein kinase family. This protein is a catalytic subunit of the highly conserved protein kinase complex known as M-phase promoting factor (MPF), which is essential for G1/S and G2/M phase transitions of eukaryotic cell cycle. Mitotic cyclins stably associate with this protein and function as regulatory subunits. The kinase activity of this protein is controlled by cyclin accumulation and destruction through the cell cycle. The phosphorylation and dephosphorylation of this protein also play important regulatory roles in cell cycle control. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
CDC2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CDC2. | |
In PBS (0.09% sodium azide) | |
NP_001777;P06493 | |
CDC2 | |
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S39 of human CDC2. | |
400 μL | |
Primary | |
Human | |
Liquid |
Dot Blot, ELISA, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
CDC2 | |
CDC28A/CDK1/DKFZp686L20222/MGC111195 | |
Rabbit | |
Protein G purification | |
RUO | |
983 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only