Learn More
CDC25A (phospho S293), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CDC25A.
Supplier: Abnova Corporation PAB0428
Description
CDC25A is a member of the CDC25 family of phosphatases. CDC25A is required for progression from G1 to the S phase of the cell cycle. It activates the cyclin-dependent kinase CDC2 by removing two phosphate groups. CDC25A is specifically degraded in response to DNA damage, which prevents cells with chromosomal abnormalities from progressing through cell division. CDC25A is an oncogene, although its exact role in oncogenesis has not been demonstrated. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
CDC25A | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CDC25A. | |
In PBS (0.09% sodium azide) | |
NP_001780;P30304 | |
CDC25A | |
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S293 of human CDC25A. | |
400 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
CDC25A | |
CDC25A2 | |
Rabbit | |
Protein G purification | |
RUO | |
993 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only