Learn More
CDC27 Rabbit anti-Chicken, Chimpanzee, Dog, Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CDC27.
Supplier: Abnova Corporation PAB9986
Description
The protein encoded by this gene shares strong similarity with Saccharomyces cerevisiae protein Cdc27, and the gene product of Schizosaccharomyces pombe nuc 2. This protein is a component of anaphase-promoting complex (APC), which is composed of eight protein subunits and highly conserved in eucaryotic cells. APC catalyzes the formation of cyclin B-ubiquitin conjugate that is responsible for the ubiquitin-mediated proteolysis of B-type cyclins. This protein and 3 other members of the APC complex contain the TPR (tetratricopeptide repeat), a protein domain important for protein-protein interaction. This protein was shown to interact with mitotic checkpoint proteins including Mad2, p55CDC and BUBR1, and thus may be involved in controlling the timing of mitosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
CDC27 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of CDC27. | |
In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
P30260;NP_001247 | |
CDC27 | |
A synthetic peptide corresponding to amino acids 422-430 of human CDC27. | |
RUO | |
996 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunofluorescence, Western Blot | |
Unconjugated | |
ELISA (1:5000-1:40000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
CDC27 | |
APC3/CDC27Hs/D0S1430E/D17S978E/HNUC | |
Rabbit | |
100 μg | |
Primary | |
Canine, Chicken, Chimpanzee, Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only