Learn More
CDC42EP5 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CDC42EP5.
Supplier: Abnova Corporation PAB28119
Description
CDC42, a small Rho GTPase, regulates the formation of F-actin-containing structures through its interaction with the downstream effector proteins. The protein encoded by this gene is a member of the Borg family of CDC42 effector proteins. Borg family proteins contain a CRIB (Cdc42/Rac interactive-binding) domain. They bind to, and negatively regulate the function of, CDC42. The specific function of this protein has not yet been determined. [provided by RefSeq
Sequence: KPDAEPRPGTQPPQARCRPNADLELNDVIGLSpecifications
| CDC42EP5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CDC42EP5. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| Q6NZY7 | |
| CDC42EP5 | |
| Recombinant protein corresponding to amino acids of recombinant CDC42EP5. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| CDC42EP5 | |
| Borg3/CEP5/MGC21945/MGC71153 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 148170 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only