Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDH1/CDH2/CDH3/CDH4, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89128707 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-707 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-707 Supplier Abnova Corporation Supplier No. PAB27057
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CDH1/CDH2/CDH3/CDH4.

Specifications

Antigen CDH1/CDH2/CDH3/CDH4
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CDH1/CDH2/CDH3/CDH4.
Dilution Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.05% sodium azide)
Gene Accession No. P12830 (Gene ID : 999);P19022 (Gene ID : 1000);P22223 (Gene ID : 1001);P55283 (Gene ID : 1002)
Gene Alias Arc-1/CD324/CDHE/ECAD/LCAM/UVO
Gene Symbols CDH1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human CDH1/CDH2/CDH3/CDH4.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 999/1000/1001/1002
Test Specificity CDH1/CDH2/CDH3/CDH4 polyclonal antibody detects endogenous levels of CDH1/CDH2/CDH3/CDH4 protein.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.