Learn More
CDK11B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CDK11B.
Supplier: Abnova Corporation PAB17355
Description
This gene encodes a member of the p34Cdc2 protein kinase family. p34Cdc2 kinase family members are known to be essential for eukaryotic cell cycle control. This gene is in close proximity to CDC2L2, a nearly identical gene in the same chromosomal region. The gene loci including this gene, CDC2L2, as well as metalloprotease MMP21/22, consist of two identical, tandemly linked genomic regions which are thought to be a part of the larger region that has been duplicated. This gene and CDC2L2 were shown to be deleted or altered frequently in neuroblastoma with amplified MYCN genes. The protein kinase encoded by this gene could be cleaved by caspases and was demonstrated to play roles in cell apoptosis. Several alternatively spliced variants of this gene have been reported. [provided by RefSeq
Specifications
CDK11B | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of CDK11B. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
P21127 | |
CDC2L1 | |
A synthetic peptide corresponding to N-terminus of human CDK11B. | |
RUO | |
984 | |
Human, Mouse | |
Liquid |
Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) ELISA (1:20000) The optimal working dilution should be determined by the end user. | |
CDC2L1 | |
CDC2L2/CDK11/CDK11-p110/CDK11-p46/CDK11-p58/CLK-1/FLJ59152/PK58/p58/p58CDC2L1/p58CLK-1 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total CDK11B protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only