Learn More
CDK2 (phospho), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CDK2.
Supplier: Abnova Corporation PAB10150
Description
The protein encoded by this gene is a member of the Ser/Thr protein kinase family. This protein kinase is highly similar to the gene products of S. cerevisiae cdc28, and S. pombe cdc2. It is a catalytic subunit of the cyclin-dependent protein kinase complex, whose activity is restricted to the G1-S phase, and essential for cell cycle G1/S phase transition. This protein associates with and regulated by the regulatory subunits of the complex including cyclin A or E, CDK inhibitor p21Cip1 (CDKN1A) and p27Kip1 (CDKN1B). Its activity is also regulated by its protein phosphorylation. Two alternatively spliced variants and multiple transcription initiation sites of this gene have been reported. [provided by RefSeq
Specifications
CDK2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CDK2. | |
In PBS (0.08% sodium azide) | |
p33(CDK2) | |
Rabbit | |
20 μg | |
Primary | |
Human | |
Liquid |
Western Blot | |
Unconjugated | |
Western blot (1:2000) The optimal working dilution should be determined by the end user. | |
CDK2 | |
CDK2 | |
Synthetic phosphopeptide corresponding to residues surrounding phosphorylated human CDK2. | |
RUO | |
1017 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only