Learn More
CDK7 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CDK7.
Supplier: Abnova Corporation PAB27106
Description
The protein encoded by this gene is a member of the cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This protein forms a trimeric complex with cyclin H and MAT1, which functions as a Cdk-activating kinase (CAK). It is an essential component of the transcription factor TFIIH, that is involved in transcription initiation and DNA repair. This protein is thought to serve as a direct link between the regulation of transcription and the cell cycle. [provided by RefSeq
Specifications
CDK7 | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
CDK7 | |
CAK1/CDKN7/MO15/STK1/p39MO15 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
1022 | |
Human, Mouse | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of CDK7. | |
In PBS, pH 7.2 (0.05% sodium azide) | |
P50613 | |
CDK7 | |
A synthetic peptide corresponding to human CDK7. | |
100 μL | |
Primary | |
CDK7 polyclonal antibody detects endogenous levels of CDK7 protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only