Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDKL2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CDKL2.
Supplier: Abnova Corporation PAB2667
Description
This gene product is a member of a large family of CDC2-related serine/threonine protein kinases. It accumulates primarily in the cytoplasm, with lower levels in the nucleus. [provided by RefSeq
Specifications
CDKL2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of CDKL2. | |
In PBS (0.09% sodium azide) | |
NP_003939;Q92772 | |
CDKL2 | |
A synthetic peptide (conjugated with KLH) corresponding to internal region of human CDKL2. | |
400 μL | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
CDKL2 | |
KKIAMRE/P56 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
8999 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction