Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDKN1A (phospho T57), Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89114273 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-273 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-273 Supplier Abnova Corporation Supplier No. PAB0556
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of CDKN1A.

This gene encodes a potent cyclin-dependent kinase inhibitor. The encoded protein binds to and inhibits the activity of cyclin-CDK2 or -CDK4 complexes, and thus functions as a regulator of cell cycle progression at G1. The expression of this gene is tightly controlled by the tumor suppressor protein p53, through which this protein mediates the p53-dependent cell cycle G1 phase arrest in response to a variety of stress stimuli. This protein can interact with proliferating cell nuclear antigen (PCNA), a DNA polymerase accessory factor, and plays a regulatory role in S phase DNA replication and DNA damage repair. This protein was reported to be specifically cleaved by CASP3-like caspases, which thus leads to a dramatic activation of CDK2, and may be instrumental in the execution of apoptosis following caspase activation. Two alternatively spliced variants, which encode an identical protein, have been reported. [provided by RefSeq

Specifications

Antigen CDKN1A
Applications Dot Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of CDKN1A.
Dilution Dot Blot (1:500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene CDKN1A
Gene Accession No. NP_000380;P38936
Gene Alias CAP20/CDKN1/CIP1/MDA-6/P21/SDI1/WAF1/p21CIP1
Gene Symbols CDKN1A
Host Species Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T57 of human CDKN1A.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1026
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.