Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDKN1B (phospho T187), Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89114277 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-277 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-277 Supplier Abnova Corporation Supplier No. PAB0560
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of CDKN1B.

This gene encodes a cyclin-dependent kinase inhibitor, which shares a limited similarity with CDK inhibitor CDKN1A/p21. The encoded protein binds to and prevents the activation of cyclin E-CDK2 or cyclin D-CDK4 complexes, and thus controls the cell cycle progression at G1. The degradation of this protein, which is triggered by its CDK dependent phosphorylation and subsequent ubiquitination by SCF complexes, is required for the cellular transition from quiescence to the proliferative state. [provided by RefSeq

Specifications

Antigen CDKN1B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of CDKN1B.
Dilution Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene CDKN1B
Gene Accession No. NP_004055;P46527
Gene Alias CDKN4/KIP1/MEN1B/MEN4/P27KIP1
Gene Symbols CDKN1B
Host Species Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T187 of human CDKN1B.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1027
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.