Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDKN1C Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89127250 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-250 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-250 Supplier Abnova Corporation Supplier No. PAB25634
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CDKN1C.

The protein encoded by this gene is a tight-binding, strong inhibitor of several G1 cyclin/Cdk complexes and a negative regulator of cell proliferation. Mutations in this gene are implicated in sporadic cancers and Beckwith-Wiedemann syndorome, suggesting that this gene is a tumor suppressor candidate. Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen CDKN1C
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CDKN1C.
Dilution Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene CDKN1C
Gene Accession No. NP_000067
Gene Alias BWCR/BWS/KIP2/WBS/p57
Gene Symbols CDKN1C
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 17 amino acids at N-terminus of human CDKN1C.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1028
Test Specificity At least three isoforms of CDKN1C are known to exist; this antibody will detect all three isoforms. Despite its predicted molecular weight, CDKN1C often migrates at 57kDa in SDS-PAGE. CDKN1C antibody is predicted to not cross react with other CIP/KIP proteins.
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.