Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDKN2A Rabbit anti-Human, DyLight 488, Polyclonal Antibody, Abnova™

Catalog No. 89117585 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-585 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-585 Supplier Abnova Corporation Supplier No. PAB15355
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CDKN2A.

This gene generates several transcript variants which differ in their first exons. At least three alternatively spliced variants encoding distinct proteins have been reported, two of which encode structurally related isoforms known to function as inhibitors of CDK4 kinase. The remaining transcript includes an alternate first exon located 20kb upstream of the remainder of the gene; this transcript contains an alternate open reading frame (ARF) that specifies a protein which is structurally unrelated to the products of the other variants. This ARF product functions as a stabilizer of the tumor suppressor protein p53 as it can interact with, and sequester, MDM1, a protein responsible for the degradation of p53. In spite of the structural and functional differences, the CDK inhibitor isoforms and the ARF product encoded by this gene, through the regulatory roles of CDK4 and p53 in cell cycle G1 progression, share a common functionality in cell cycle G1 control. This gene is frequently mutated or deleted in a wide variety of tumors, and is known to be an important tumor suppressor gene. [provided by RefSeq

Specifications

Antigen CDKN2A
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate DyLight 488
Description Rabbit polyclonal antibody raised against synthetic peptide of CDKN2A.
Dilution Immunofluorescence (1:100) Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium borate
Gene CDKN2A
Gene Accession No. Q8N726
Gene Alias ARF/CDK4I/CDKN2/CMM2/INK4/INK4a/MLM/MTS1/TP16/p14/p14ARF/p16/p16INK4/p16INK4a/p19
Gene Symbols CDKN2A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 50-150 of human CDKN2A.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1029
Test Specificity This antibody reacts with human p14 ARF. Other species untested. This is specific to p14ARF. This antibody may be used for Western blotting, where a band is seen at where a band is seen at ∼16 KDa, representing p14ARF (p14ARF tends to run slightly higher than the theoretical MW of 14 KDa). Additional faint bands may be seen at ∼32 and 47 KDa.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.