Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdkn2a, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115644 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-644 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-644 Supplier Abnova Corporation Supplier No. PAB12146
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Cdkn2a.

Specifications

Antigen Cdkn2a
Applications ELISA, Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Cdkn2a.
Dilution Western Blot (2 ug/mL) Immunocytochemistry (1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide)
Gene Cdkn2a
Gene Accession No. Q64364;P51480
Gene Alias ARF-INK4a/Arf/INK4a-ARF/Ink4a/Arf/MTS1/Pctr1/p16/p16(INK4a)/p16INK4a/p19/p19ARF
Gene Symbols Cdkn2a
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 50-150 of mouse Cdkn2a.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12578
Test Specificity On Western blot This antibody detects a band corresponding to p19ARF in mouse embryonic fibroblasts.
Target Species Mouse
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.