Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEBPD, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89127304 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-304 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-304 Supplier Abnova Corporation Supplier No. PAB25683
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CEBPD.

The protein encoded by this intronless gene is a bZIP transcription factor which can bind as a homodimer to certain DNA regulatory regions. It can also form heterodimers with the related protein CEBP-alpha. The encoded protein is important in the regulation of genes involved in immune and inflammatory responses, and may be involved in the regulation of genes associated with activation and/or differentiation of macrophages. [provided by RefSeq

Specifications

Antigen CEBPD
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CEBPD.
Dilution Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene CEBPD
Gene Accession No. NP_005186
Gene Alias C/EBP-delta/CELF/CRP3/NF-IL6-beta
Gene Symbols CEBPD
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 18 amino acids at C-terminus of human CEBPD.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1052
Test Specificity Multiple transcript variants encoding different isoforms have been found for this gene.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.