Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cebpd, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114682 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-682 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-682 Supplier Abnova Corporation Supplier No. PAB10182
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Cebpd.

Specifications

Antigen Cebpd
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Cebpd.
Dilution ELISA (1:4000-1:9000) Western Blot (1:1000-1:15000) Immunoprecipitation (0.8-1.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene Cebpd
Gene Accession No. NP_031705;Q00322
Gene Symbols Cebpd
Host Species Rabbit
Immunogen Recombinant protein corresponding to N-terminus of mouse Cebpd.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12609
Test Specificity No cross-reactivity is observed with mouse C/EBPalpha or C/EBPbeta.
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.