Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Hemolysin, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137459 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-459 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-459 Supplier Abnova Corporation Supplier No. PAB11352
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against Cell Hemolysin.

Specifications

Antigen Cell Hemolysin
Applications Agglutination
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Cell Hemolysin.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from antiserum
Host Species Rabbit
Immunogen Native from sheep washed pooled red blood cells (RBC).
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Sheep
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 2mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.