Learn More
CELSR2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CELSR2.
Supplier: Abnova Corporation PAB17504
Description
The protein encoded by this gene is a member of the flamingo subfamily, part of the cadherin superfamily. The flamingo subfamily consists of nonclassic-type cadherins; a subpopulation that does not interact with catenins. The flamingo cadherins are located at the plasma membrane and have nine cadherin domains, seven epidermal growth factor-like repeats and two laminin A G-type repeats in their ectodomain. They also have seven transmembrane domains, a characteristic unique to this subfamily. It is postulated that these proteins are receptors involved in contact-mediated communication, with cadherin domains acting as homophilic binding regions and the EGF-like domains involved in cell adhesion and receptor-ligand interactions. The specific function of this particular member has not been determined. [provided by RefSeq
Specifications
CELSR2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of CELSR2. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
Q9HCU4 | |
CELSR2 | |
A synthetic peptide corresponding to internal of human CELSR2. | |
RUO | |
1952 | |
Human, Mouse, Rat | |
Liquid |
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
CELSR2 | |
CDHF10/EGFL2/FLJ34118/FLJ42737/FLJ45143/FLJ45845/Flamingo1/KIAA0279/MEGF3 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total CELSR2 protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only