Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENPF, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137491 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-491 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-491 Supplier Abnova Corporation Supplier No. PAB12373
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant CENPF.

Specifications

Antigen CENPF
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant CENPF.
Dilution Flow Cytometry (1:50-1:200) Immunocytochemistry (1:200-1:1000) Immunofluorescence (1:750) Immunoprecipitation (1:50) Western Blot (1:1500-1:2500) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 2.0mg/mL BSA, 0.05% sodium azide)
Gene Accession No. P49454
Host Species Rabbit
Immunogen Recombinant protein corresponding to C-terminus of CENPF.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific to CENP-F.
Target Species Human
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.