Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP85L Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89123958 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-958 Supplier Abnova Corporation Supplier No. PAB22252
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant CEP85L.

Sequence: SKLRESLTPDGSKWSTSLMQTLGNHSRGEQDSSLDMKDFRPLRKWSSLSKLTAPDNCGQGGTVCREESRNGLEK

Specifications

Antigen CEP85L
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant CEP85L.
Dilution Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene CEP85L
Gene Accession No. Q5SZL2
Gene Alias RP11-57K17.2/C6orf204/NY-BR-15/bA57K17.2
Gene Symbols CEP85L
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human CEP85L.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387119
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.