Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERK Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89118193 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-193 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-193 Supplier Abnova Corporation Supplier No. PAB16321
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CERK.

CERK converts ceramide to ceramide 1-phosphate (C1P), a sphingolipid metabolite. Both CERK and C1P have been implicated in various cellular processes, including proliferation, apoptosis, phagocytosis, and inflammation (Kim et al., 2006 [PubMed 16488390]).[supplied by OMIM

Specifications

Antigen CERK
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CERK.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene CERK
Gene Accession No. Q8TCT0
Gene Alias DKFZp434E0211/FLJ21430/FLJ23239/KIAA1646/LK4/MGC131878/dA59H18.2/dA59H18.3/hCERK
Gene Symbols CERK
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human CERK.
Purification Method Immunoaffinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64781
Test Specificity N-terminal extracellular domain of human.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.