Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CESA1 Rabbit anti-Thale cress, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CESA1.
Supplier: Abnova Corporation PAB16055
Specifications
CESA1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of CESA1. | |
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
CELLULOSE SYNTHASE 1/F8B4.110/F8B4_110/RADIALLY SWOLLEN 1/RSW1 | |
Rabbit | |
100 μg | |
Primary | |
Thale cress | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
CESA1 | |
CESA1 | |
A synthetic peptide corresponding to N-terminus of Arabidopsis thaliana CESA1. | |
RUO | |
829376 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction