Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CFL1/CFL2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89128479 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-479 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-479 Supplier Abnova Corporation Supplier No. PAB26834
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CFL1/CFL2.

Sequence: A-T-Yp-E-T

Specifications

Antigen CFL1/CFL2
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CFL1/CFL2.
Dilution Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
Gene Accession No. P23528 (Gene ID : 1072);Q9Y281 (Gene ID : 1073)
Gene Alias CFL
Gene Symbols CFL1
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to residues surrounding Y88 of human CFL1/CFL2.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1072/1073
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.