Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cflar, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115475 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-475 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-475 Supplier Abnova Corporation Supplier No. PAB11962
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Cflar.

Specifications

Antigen Cflar
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Cflar.
Dilution Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.02% sodium azide)
Gene Cflar
Gene Accession No. NP_997536
Gene Alias 2310024N18Rik/A430105C05Rik/AI646576/AU021929/CLARP/Cash/Casper/ENSMUSG00000072980/FLAME/Flip/I-FLICE/MGC103395/MRIT/c-Flip
Gene Symbols Cflar
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 449-465 of mouse Cflar.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12633
Test Specificity This antibody recognizes mouse FLIPL form (55 kD) but not FLIPS form. Epitopes at the C-terminus of mouse FLIP L form/CASH alpha form.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.