Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ch-TOG, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89138295 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-295 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-295 Supplier Abnova Corporation Supplier No. PAB17111
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ch-TOG.

Specifications

Antigen ch-TOG
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ch-TOG.
Dilution Western Blot (1:5000-1:10000) Immunoprecipitation (1:5000-1:10000) The optimal working dilution should be determined by the end user.
Formulation In antiserum
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus amino acids of ch-TOG.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes human and mouse ch-TOG. A BLAST search demonstrated 100% identity to the peptide immunogen in human, mouse, rat, chimpanzee, dog, cow, chicken, opossum, and frog.
Target Species Bovine, Canine, Chicken, Chimpanzee, Frog, Human, Mouse, Opossum, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.