Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHCHD7, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89128859 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-859 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-859 Supplier Abnova Corporation Supplier No. PAB27205
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CHCHD7.

Specifications

Antigen CHCHD7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CHCHD7.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene CHCHD7
Gene Accession No. NP_077276
Gene Alias FLJ40966/MGC2217
Gene Symbols CHCHD7
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acids at C-terminus of human CHCHD7.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79145
Test Specificity Three alternatively spliced transcript variants have been observed. CHCHD7 is predicted to not cross-react to other members of the CHCHD family.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.