Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-CHD1L, Polyclonal,

Catalog No. ABE1307MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1307MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1307MI Supplier MilliporeSigma™ Supplier No. ABE1307
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects CHD1L in Human samples, and it is validated for Western Blotting

Chromodomain helicase/ATPase DNA binding protein 1-like gene (CHD1L) is a ∼100.9kDa DNA helicase that is amplified in many solid tumors. The CHD1L gene, located at Chr 1q21 is frequently amplified in hepatocellular carcinoma. CHD1L plays a role in chromatin remodeling following DNA damage. It is targeted to DNA damage sites through interaction with poly(ADP)-ribose) and regulated chromatin during DNA repair. CHD1L contains a SNF2-N domain and a helicase superfamily domain, but unlike CHD1, it does not contain a chromodomain, but instead contains a c-terminal macrodomain, which is an (ADP)-ribose/ polymer of the ADP-ribose (PAR)-binding element. In addition to PAR binding, the macro domain of CHD1L is able to bind Nur77, a critical member of a p53-independent apoptotic pathway. This binding subsequently inhibits the nucleus-to-mitochondria translocation of Nur77 and disrupts apoptotic cell death. Overexpression of CHD1L protein in tumors is considered to be an independent biomarker of poor prognosis and short tumor-free survival time.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD1L
Applications Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q86WJ1
Gene Symbols CHD1L; ALC1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the Helicase C-terminal domain of human CHD1L.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_004275
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.