Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD5, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89116922 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-922 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-922 Supplier Abnova Corporation Supplier No. PAB14086
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CHD5.

Changes in gene expression are associated with changes in chromatin structure, which can result from modification of histones by acetylation and methylation. In addition, SWI/SNF proteins can effect chromatin structure and gene expression by causing ATP-dependent disruption of the nucleosome and by enhancing binding of transcription factors. CHD5 belongs to a group of SWI/SNF proteins called CHD proteins, which contain a SWI/SNF-like helicase/ATPase domain, as well as a DNA-binding domain and a chromodomain that directly modifies chromatin structure (Thompson et al., 2003 [PubMed 12592387]).[supplied by OMIM

Specifications

Antigen CHD5
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CHD5.
Dilution ELISA (1:500-1:1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In serum (0.05% sodium azide)
Gene CHD5
Gene Accession No. Q8TDI0
Gene Alias DKFZp434N231/KIAA0444
Gene Symbols CHD5
Host Species Rabbit
Immunogen A mixture of synthetic peptides (conjugated with KLH) that corresponding N-terminus and to internal region of human CHD5.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26038
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.