Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEK1 Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89118799 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-799 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-799 Supplier Abnova Corporation Supplier No. PAB17001
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CHEK1.

Specifications

Antigen CHEK1
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CHEK1.
Dilution Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:20000) The optimal working dilution should be determined by the end user.
Formulation In 20mM PBS, 0.15M NaCl, pH 7.2 (0.01% sodium azide)
Gene CHEK1
Gene Accession No. O14757
Gene Alias CHK1
Gene Symbols CHEK1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues surrounding S317 of human CHEK1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1111
Test Specificity This antibody detects endogenous levels of total CHEK1 protein.
Target Species Human, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.