Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHM, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121689 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-689 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-689 Supplier Abnova Corporation Supplier No. PAB20184
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant CHM.

This gene encodes component A of the RAB geranylgeranyl transferase holoenzyme. In the dimeric holoenzyme, this subunit binds unprenylated Rab GTPases and then presents them to the catalytic Rab GGTase subunit for the geranylgeranyl transfer reaction. Rab GTPases need to be geranylgeranyled on either one or two cysteine residues in their C-terminus to localize to the correct intracellular membrane. Mutations in this gene are a cause of choroideremia; also known as tapetochoroidal dystrophy (TCD). This X-linked disease is characterized by progressive dystrophy of the choroid, retinal pigment epithelium and retina. Alternative splicing results in multiple transcript variants encoding different isoforms

Sequence: SRKDKTIQHVEVFCYASQDLHEDVEEAGALQKNHALVTSANSTEAADSAFLPTEDESLSTMSCEMLTEQTPSSDPENALEVNGAEVTGEKENHCDDKTCVPSTSAEDMSENVPIAEDTTEQPKKNRITYSQIIKEG

Specifications

Antigen CHM
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant CHM.
Dilution Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene CHM
Gene Accession No. P24386
Gene Alias DXS540/FLJ38564/GGTA/HSD-32/MGC102710/REP-1/TCD
Gene Symbols CHM
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human CHM.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1121
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.