Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHST10, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89119309 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-309 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-309 Supplier Abnova Corporation Supplier No. PAB17645
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CHST10.

Cell surface carbohydrates modulate a variety of cellular functions and are typically synthesized in a stepwise manner. HNK1ST plays a role in the biosynthesis of HNK1 (see MIM 151290), a neuronally expressed carbohydrate that contains a sulfoglucuronyl residue.[supplied by OMIM

Specifications

Antigen CHST10
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CHST10.
Dilution Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:40000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Gene CHST10
Gene Accession No. O43529
Gene Alias HNK-1ST/HNK1ST/MGC17148
Gene Symbols CHST10
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal of human CHST10.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9486
Test Specificity This antibody detects endogenous levels of total CHST10 protein.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.