Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHUK Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89116648 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-648 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-648 Supplier Abnova Corporation Supplier No. PAB13402
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CHUK.

This gene encodes a member of the serine/threonine protein kinase family. The encoded protein, a component of a cytokine-activated protein complex that is an inhibitor of the essential transcription factor NF-kappa-B complex, phosphorylates sites that trigger the degradation of the inhibitor via the ubiquination pathway, thereby activating the transcription factor. [provided by RefSeq

Specifications

Antigen CHUK
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CHUK.
Dilution Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene CHUK
Gene Accession No. AF009225
Gene Alias IKBKA/IKK-alpha/IKK1/IKKA/NFKBIKA/TCF16
Gene Symbols CHUK
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 658-674 of human CHUK.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1147
Test Specificity human IKK alpha differs from corresponding murine sequence by one amino acid. An 85 KDa band should be detected. This polyclonal antibody has no cross response to IKKb or IKKy.
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.