Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIB2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89131247 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-131-247 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-131-247 Supplier Abnova Corporation Supplier No. PAB4036
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CIB2.

The amino acid sequence the protein encoded by this gene is similar to that of KIP/CIB, calcineurin B, and calmodulin. This suggests that the encoded protein may be a Ca2+-binding regulatory protein that interacts with DNA-dependent protein kinase catalytic subunit (DNA-PKcs). [provided by RefSeq]

Specifications

Antigen CIB2
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CIB2.
Dilution ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene CIB2
Gene Accession No. O75838
Gene Alias KIP2
Gene Symbols CIB2
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human CIB2.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10518
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.