Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIC Rabbit anti-Dog, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89115831 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-831 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-831 Supplier Abnova Corporation Supplier No. PAB12457
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant CIC.

Specifications

Antigen CIC
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant CIC.
Dilution Immunoprecipitation (2-5 ug/mg lysate) The optimal working dilution should be determined by the end user.
Formulation In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide)
Gene CIC
Gene Accession No. Q96RK0
Gene Alias KIAA0306
Gene Symbols CIC
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 1150-1200 of human CIC.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23152
Target Species Canine, Human, Mouse, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.