Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cidea, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115473 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-473 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-473 Supplier Abnova Corporation Supplier No. PAB11960
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Cidea.

Specifications

Antigen Cidea
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Cidea.
Dilution Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.02% sodium azide)
Gene Cidea
Gene Accession No. AAC34985
Gene Alias AW212747
Gene Symbols Cidea
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 200-214 of mouse Cidea.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12683
Test Specificity This antibody recognizes mouse CIDE-A (25kD). No cross-reactivity with CIDE-B. Epitopes at the C-terminus of CIDE-A.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.