Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-CKLF, Polyclonal, Abnova™

Catalog No. 89117169 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-169 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-169 Supplier Abnova Corporation Supplier No. PAB14594

Specifications

Antigen CKLF
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS (0.2% gelatin, 0.05% sodium azide)
Gene Accession No. NP_001035228
Gene Symbols CKLF
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human CKLF.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 51192
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.