Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLCN6 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89128629 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-629 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-629 Supplier Abnova Corporation Supplier No. PAB26982
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CLCN6.

The CLCN family of voltage-dependent chloride channel genes comprises nine members (CLCN1-7, Ka and Kb) which demonstrate quite diverse functional characteristics while sharing significant sequence homology. Chloride channel 6 and 7 belong to a subbranch of this family. Chloride channel 6 has four different alternatively spliced transcript variants. This gene is in close vicinity to two other kidney-specific chloride channel genes, CLCNKA and CLCNKB. [provided by RefSeq

Specifications

Antigen CLCN6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CLCN6.
Dilution Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Gene CLCN6
Gene Accession No. P51797
Gene Alias CLC-6/KIAA0046
Gene Symbols CLCN6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human CLCN6.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1185
Test Specificity CLCN6 polyclonal antibody detects endogenous levels of CLCN6 protein.
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.