Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLIC1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120323 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-323 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-323 Supplier Abnova Corporation Supplier No. PAB18767
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CLIC1.

Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 1 is a member of the p64 family; the protein localizes principally to the cell nucleus and exhibits both nuclear and plasma membrane chloride ion channel activity. [provided by RefSeq

Specifications

Antigen CLIC1
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CLIC1.
Dilution ELISA (1:20000-1:80000) Western Blot (1:200-1:500) Immunohistochemistry (1:50-1:250) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene CLIC1
Gene Accession No. NM_001288
Gene Alias G6/NCC27
Gene Symbols CLIC1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 14 amino acids at N-terminus of human CLIC1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1192
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.