Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabbit anti-CLN5, Polyclonal, Abnova™
Supplier: Abnova Corporation PAB24179
Specifications
CLN5 | |
Polyclonal | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
CLN5 | |
Recombinant protein corresponding to amino acids of human CLN5. | |
100 μL | |
1203 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
O75503 | |
Rabbit | |
Affinity Purified | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction