Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLTA/CLTB, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117030 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-030 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-030 Supplier Abnova Corporation Supplier No. PAB14385
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CLTA/CLTB.

Sequence: EEDPAAAFLAQQESEIAGIEND

Specifications

Antigen CLTA/CLTB
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CLTA/CLTB.
Dilution Western Blot (1:1000-1:2000) Immunofluorescence (1:100-1:500) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from serum
Gene Accession No. P09496
Gene Alias LCA
Gene Symbols CLTA
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human CLTA/CLTB.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1211/1212
Test Specificity This antibody recognizes both CLCa and CLCb isoforms of human CLC. This antibody recognizes bands migrating around 30 KDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C or -20°C on dry atmosphere. After reconstitution with water and glycerol (1:1), store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.