Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMPK2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89127227 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-227 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-227 Supplier Abnova Corporation Supplier No. PAB25613
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CMPK2.

Mitochondrial UMP-CMP kinase (EC 2.7.2.14) is a component of the salvage pathway for nucleotide synthesis. Other enzymes of the salvage pathway include thymidine kinase-2 (TK2; MIM 188250), deoxynucleotidase-2 (NT5M; MIM 605292), deoxyguanosine kinase (DGUOK; MIM 601465), adenylate kinase-2 (AK2; MIM 103020), adenylate kinase-3 (AK3; MIM 609290), adenylate kinase-3-like-1 (AK3L1; MIM 103030), and nucleoside diphosphate kinase (NME4; MIM 601818) (Xu et al., 2008 [PubMed 17999954]).[supplied by OMIM

Specifications

Antigen CMPK2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CMPK2.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene CMPK2
Gene Accession No. NP_997198
Gene Alias TYKi/UMP-CMPK2
Gene Symbols CMPK2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 19 amino acids at C-terminus of human CMPK2.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129607
Test Specificity Multiple isoforms of CMPK2 are known to exist.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.