Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CMYA5, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120975 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-975 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-975 Supplier Abnova Corporation Supplier No. PAB19440
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CMYA5.

Specifications

Antigen CMYA5
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CMYA5.
Dilution Western Blot (1-2 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene CMYA5
Gene Accession No. NP_705838
Gene Alias C5orf10/DKFZp451G182/DKFZp451G223/SPRYD2/TRIM76
Gene Symbols CMYA5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 18 amino acids near C-terminus of human CMYA5.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 202333
Test Specificity This antibody is predicted to not cross-react with other SPRYD protein family members. At least four isoforms of SPRYD2 are known to exist.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.