Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNRIP1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89128934 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-934 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-934 Supplier Abnova Corporation Supplier No. PAB27423
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CNRIP1.

This gene encodes a G-protein coupled receptor which interacts with the C-terminal tail of cannabinoid receptor 1. This receptor plays a role in synaptic plasticity, analgesia, appetite, and neuroprotection. Two transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq

Specifications

Antigen CNRIP1
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CNRIP1.
Dilution Western Blot (1-2 ug/mL) Immunohistochemistry (2.5 ug/mL) Immunofluorescence (20 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene CNRIP1
Gene Accession No. NP_056278
Gene Alias C2orf32/CRIP1/DKFZp566K1924
Gene Symbols CNRIP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acids at internal region of human CNRIP1.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25927
Test Specificity At least two isoforms of CNRIP1 are known to exist; this antibody will detect both isoforms.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.