Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cntn6 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89117003 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-003 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-003 Supplier Abnova Corporation Supplier No. PAB14323
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Cntn6.

Sequence: GYPSPHYRWKQ

Specifications

Antigen Cntn6
Applications Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Cntn6.
Dilution Immunohistochemistry (1:1000-1:4000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Cntn6
Gene Alias NB-3
Gene Symbols Cntn6
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 54-64 of mouse Cntn6.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53870
Test Specificity This antiserum stains beautifully the NB3 in DRG, where it nicely stains a subpopulation of neurons and in the trigeminal nerve root, where staining is restricted to the oligodendrocytes.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.