Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Col1a1/Col1a2 Rabbit anti-Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native Col1a1/Col1a2.
Supplier: Abnova Corporation PAB13488
Specifications
Col1a1/Col1a2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against native Col1a1/Col1a2. | |
No additive | |
Col1a1 | |
Native purified Col1a1/Col1a2 mouse skin. | |
RUO | |
12842/12843 | |
Mouse | |
Lyophilized |
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Radioimmune Assays (RIA) | |
Unconjugated | |
Immunohistochemistry (1:200-1:600) Immunofluorescence (1:40) The optimal working dilution should be determined by the end user. | |
Col1a-1/Cola-1/Cola1/Mov-13/Mov13 | |
Rabbit | |
100 μL | |
Primary | |
This antibody reacts with mouse collagen type I 100%, mouse collagen type II and IV <0.1%; mouse collage type III <1%;. | |
Store at 4°C. For long term storage store at -20°C. Aliquot after reconstitution to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction