Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL1A1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89118877 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-877 Supplier Abnova Corporation Supplier No. PAB17205
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of COL1A1.

This gene encodes the pro-alpha1 chains of type I collagen whose triple helix comprises two alpha1 chains and one alpha2 chain. Type I is a fibril-forming collagen found in most connective tissues and is abundant in bone, cornea, dermis and tendon. Mutations in this gene are associated with osteogenesis imperfecta types I-IV, Ehlers-Danlos syndrome type VIIA, Ehlers-Danlos syndrome Classical type, Caffey Disease and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene. [provided by R. Dalgleish

Specifications

Antigen COL1A1
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of COL1A1.
Dilution Western Blot (1:1000) Immunohistochemistry (1:100) The optimal working dilution should be determined by the end user.
Formulation In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 0.1mg/mL BSA)
Gene COL1A1
Gene Accession No. P02452
Gene Alias OI4
Gene Symbols COL1A1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids at C-terminus of human COL1A1.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1277
Test Specificity This antibody has been directly tested for reactivity in human, mouse, rat and sheep. Based on the homology of the immunogen this antibody is expected to recognize the collagen I 1 polypeptide in all mammals, birds, and amphibians. specific to the ∼ 140 KDa telopeptide portion of the collagen I 1 polypeptide. This antibody works well for immunohistochemistry on paraformaldehyde-fixed sections with a simple antigen-retrieval protocol (incubate slides for 20 minutes at 90°C in 10 mM sodium citrate (pH 6.0)/ 0.1% Tween-20). Note that in paraffin sections of formaldehyde-fixed fibrotic mouse lung tissue, This antibody recognizes mature collagen I that has formed fibrils in the extracellular matrix.
Target Species Avian, Human, Mammalia, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.